Assay Details
Gene Symbol: Rqcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507611.2 | XP_003507659.1 | 2 - 3 | 584 | 79 | |
GenBank mRNA | JI873216.1 | - | 2 - 3 | 305 | 79 | |
JP055892.1 | - | 2 - 3 | 305 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
