Assay Details
Gene Symbol: Spg20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502949.2 | XP_003502997.1 | 6 - 7 | 2036 | 65 | |
XM_007643555.1 | XP_007641745.1 | 7 - 8 | 1696 | 65 | ||
XM_007643556.1 | XP_007641746.1 | 6 - 7 | 1619 | 65 | ||
GenBank mRNA | JI880013.1 | - | 2 - 3 | 251 | 65 | |
JP057739.1 | - | 2 - 3 | 251 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
