Assay Details
| Gene Symbol: Sgcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509718.2 | XP_003509766.1 | 4 - 5 | 417 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sgcd| Gene Symbol: Sgcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509718.2 | XP_003509766.1 | 4 - 5 | 417 | 75 | |
Set Membership: |
|