Assay Details
| Gene Symbol: Cpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495731.2 | XP_003495779.1 | 1 - 2 | 307 | 66 | |
| GenBank mRNA | JI867442.1 | - | 1 - 2 | 141 | 66 | |
| JP057752.1 | - | 1 - 2 | 141 | 66 | ||
More Information
Set Membership: |
|

Cpt2
Genomic Map