Assay Details
| Gene Symbol: Ahsg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508220.1 | XP_003508268.1 | 6 - 7 | 762 | 90 | |
| XM_007648843.1 | XP_007647033.1 | 6 - 7 | 741 | 90 | ||
| XM_007648844.1 | XP_007647034.1 | 3 - 4 | 360 | 90 | ||
More Information
Set Membership: |
|

Ahsg
Genomic Map