Assay Details
| Gene Symbol: Mbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507354.2 | XP_003507402.1 | 2 - 3 | 420 | 70 | |
| XM_007647936.1 | XP_007646126.1 | 2 - 3 | 747 | 70 | ||
More Information
Set Membership: |
|

Mbd4
Genomic Map