Assay Details
| Gene Symbol: LOC103160839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007620087.1 | XP_007618277.1 | 1 | 824 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103160839| Gene Symbol: LOC103160839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007620087.1 | XP_007618277.1 | 1 | 824 | 91 | |
Set Membership: |
|