Assay Details
| Gene Symbol: Glra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508680.2 | XP_003508728.1 | 1 - 2 | 75 | 63 | |
| XM_007649306.1 | XP_007647496.1 | 1 - 2 | 75 | 63 | ||
More Information
Set Membership: |
|

Glra3
Genomic Map