Assay Details
| Gene Symbol: Gabrd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509283.2 | XP_003509331.2 | 8 - 9 | 1093 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabrd| Gene Symbol: Gabrd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509283.2 | XP_003509331.2 | 8 - 9 | 1093 | 60 | |
Set Membership: |
|