Assay Details
| Gene Symbol: Flvcr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502785.2 | XP_003502833.1 | 8 - 9 | 1911 | 59 | |
| XM_007643399.1 | XP_007641589.1 | 9 - 10 | 1901 | 59 | ||
More Information
Set Membership: |
|

Flvcr2
Genomic Map