Assay Details
| Gene Symbol: Spred2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498101.2 | XP_003498149.1 | 5 - 6 | 698 | 66 | |
| GenBank mRNA | JI870849.1 | - | 5 - 6 | 632 | 66 | |
| JP060944.1 | - | 5 - 6 | 632 | 66 | ||
More Information
Set Membership: |
|

Spred2
Genomic Map