Assay Details
| Gene Symbol: Lyrm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652454.1 | XP_007650644.1 | 3 - 4 | 458 | 71 | |
| XM_007652455.1 | XP_007650645.1 | 3 - 4 | 452 | 71 | ||
| XM_007652456.1 | XP_007650646.1 | 3 - 4 | 442 | 71 | ||
| XM_007652457.1 | XP_007650647.1 | 2 - 3 | 277 | 71 | ||
| XM_007652458.1 | XP_007650648.1 | 4 - 5 | 454 | 71 | ||
| XM_007652459.1 | XP_007650649.1 | 4 - 5 | 409 | 71 | ||
| XM_007652460.1 | XP_007650650.1 | 4 - 5 | 481 | 71 | ||
| GenBank mRNA | JI888886.1 | - | 4 - 5 | 417 | 71 | |
More Information
Set Membership: |
|

Lyrm1
Genomic Map