Assay Details
| Gene Symbol: Snai2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505903.2 | XP_003505951.1 | 1 - 2 | 437 | 98 | |
| GenBank mRNA | JI874295.1 | - | 2 - 3 | 98 | ||
| JP059619.1 | - | 1 - 2 | 108 | 98 | ||
More Information
Set Membership: |
|

Snai2
Genomic Map