Assay Details
| Gene Symbol: Art5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509702.1 | XP_003509750.1 | 2 - 3 | 789 | 60 | |
| XM_007650341.1 | XP_007648531.1 | 2 - 3 | 789 | 60 | ||
More Information
Set Membership: |
|

Art5
Genomic Map