Assay Details
Gene Symbol: Rbak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496281.2 | XP_003496329.1 | 4 - 5 | 1322 | 116 | |
XM_007648770.1 | XP_007646960.1 | 4 - 5 | 521 | 116 | ||
XM_007648775.1 | XP_007646965.1 | 4 - 5 | 1341 | 116 | ||
XM_007648780.1 | XP_007646970.1 | 4 - 5 | 316 | 116 | ||
XM_007648787.1 | XP_007646977.1 | 4 - 5 | 532 | 116 | ||
GenBank mRNA | JI862023.1 | - | 1 - 2 | 93 | 116 | |
JP054536.1 | - | 1 - 2 | 93 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
