Assay Details
| Gene Symbol: Upk3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641686.1 | XP_007639876.1 | 2 - 3 | 237 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Upk3b| Gene Symbol: Upk3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641686.1 | XP_007639876.1 | 2 - 3 | 237 | 58 | |
Set Membership: |
|