Assay Details
| Gene Symbol: Star | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498222.2 | XP_003498270.1 | 3 - 4 | 859 | 66 | |
| XM_007624588.1 | XP_007622778.1 | 3 - 4 | 860 | 66 | ||
More Information
Set Membership: |
|

Star
Genomic Map