Assay Details
Gene Symbol: Nsun5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641663.1 | XP_007639853.1 | 7 - 8 | 1181 | 62 | |
GenBank mRNA | JI890278.1 | - | 1 - 2 | 62 | ||
JP057461.1 | - | 1 - 2 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
