Assay Details
| Gene Symbol: Uvssa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504111.2 | XP_003504159.1 | 13 - 14 | 2194 | 65 | |
| XM_007644632.1 | XP_007642822.1 | 13 - 14 | 2068 | 65 | ||
| XM_007644633.1 | XP_007642823.1 | 13 - 14 | 1919 | 65 | ||
| GenBank mRNA | JI874853.1 | - | 2 - 3 | 286 | 65 | |
| JP047544.1 | - | 2 - 3 | 286 | 65 | ||
More Information
Set Membership: |
|

Uvssa
Genomic Map