Assay Details
Gene Symbol: Ndufv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504441.2 | XP_003504489.1 | 2 - 3 | 247 | 62 | |
GenBank mRNA | JI889204.1 | - | 2 - 3 | 234 | 62 | |
JP056576.1 | - | 2 - 3 | 234 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
