Assay Details
Gene Symbol: Agrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508551.2 | XP_003508599.1 | 3 - 4 | 628 | 66 | |
XM_007649178.1 | XP_007647368.1 | 2 - 3 | 784 | 66 | ||
XM_007649179.1 | XP_007647369.1 | 2 - 3 | 546 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
