Assay Details
| Gene Symbol: Tmub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501295.2 | XP_003501343.1 | 1 - 2 | 1148 | 76 | |
| XM_007641966.1 | XP_007640156.1 | 2 - 3 | 797 | 76 | ||
| XM_007641967.1 | XP_007640157.1 | 2 - 3 | 1022 | 76 | ||
| XM_007641968.1 | XP_007640158.1 | 2 - 3 | 1026 | 76 | ||
| XM_007641969.1 | XP_007640159.1 | 2 - 3 | 500 | 76 | ||
| XM_007641970.1 | XP_007640160.1 | 2 - 3 | 550 | 76 | ||
| GenBank mRNA | JP063512.1 | - | 1 - 2 | 416 | 76 | |
More Information
Set Membership: |
|

Tmub1
Genomic Map