Assay Details
Gene Symbol: Ccdc132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496953.2 | XP_003497001.1 | 27 - 28 | 2892 | 58 | |
GenBank mRNA | JI895861.1 | - | 21 - 22 | 2298 | 58 | |
JP042473.1 | - | 21 - 22 | 2298 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
