Assay Details
| Gene Symbol: Tal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495859.2 | XP_003495907.1 | 1 | 221 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tal2| Gene Symbol: Tal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495859.2 | XP_003495907.1 | 1 | 221 | 92 | |
Set Membership: |
|