Assay Details
| Gene Symbol: Tmem38b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644679.1 | XP_007642869.1 | 4 - 5 | 633 | 68 | |
| GenBank mRNA | JP046857.1 | - | 3 - 4 | 429 | 68 | |
More Information
Set Membership: |
|

Tmem38b
Genomic Map