Assay Details
| Gene Symbol: Irx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007655183.1 | XP_007653373.1 | 2 - 3 | 162 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Irx4| Gene Symbol: Irx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007655183.1 | XP_007653373.1 | 2 - 3 | 162 | 58 | |
Set Membership: |
|