Assay Details
| Gene Symbol: Ccdc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651160.1 | XP_007649350.1 | 2 - 3 | 700 | 93 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc8| Gene Symbol: Ccdc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651160.1 | XP_007649350.1 | 2 - 3 | 700 | 93 | |