Assay Details
Gene Symbol: Pts | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498538.2 | XP_003498586.1 | 2 - 3 | 237 | 64 | |
GenBank mRNA | JP049899.1 | - | 2 - 3 | 236 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
