Assay Details
Gene Symbol: Rab11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510502.2 | XP_003510550.1 | 1 - 2 | 135 | 87 | |
GenBank mRNA | JI895582.1 | - | 1 - 2 | 99 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
