Assay Details
| Gene Symbol: Nkx2-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650520.1 | XP_007648710.1 | 9 - 10 | 1498 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkx2-2| Gene Symbol: Nkx2-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650520.1 | XP_007648710.1 | 9 - 10 | 1498 | 93 | |
Set Membership: |
|