Assay Details
| Gene Symbol: Slc22a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505935.1 | XP_003505983.1 | 8 - 9 | 1416 | 67 | |
| GenBank mRNA | JI879909.1 | - | 4 - 5 | 574 | 67 | |
More Information
Set Membership: |
|

Slc22a14
Genomic Map