Assay Details
| Gene Symbol: Mrpl53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511134.2 | XP_003511182.1 | 1 - 2 | 449 | 87 | |
| GenBank mRNA | JI890235.1 | - | 1 - 2 | 116 | 87 | |
More Information
Set Membership: |
|

Mrpl53
Genomic Map