Assay Details
| Gene Symbol: Mrpl53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511134.2 | XP_003511182.1 | 2 - 3 | 564 | 113 | |
| GenBank mRNA | JI890235.1 | - | 2 - 3 | 231 | 113 | |
More Information
Set Membership: |
|

Mrpl53
Genomic Map