Assay Details
| Gene Symbol: Flt3lg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649286.1 | XP_007647476.1 | 4 - 5 | 590 | 72 | |
| XM_007649287.1 | XP_007647477.1 | 4 - 5 | 572 | 72 | ||
| XM_007649288.1 | XP_007647478.1 | 3 - 4 | 536 | 72 | ||
| XM_007649289.1 | XP_007647479.1 | 5 - 6 | 448 | 72 | ||
| XM_007649290.1 | XP_007647480.1 | 4 - 5 | 590 | 72 | ||
| XM_007649291.1 | XP_007647481.1 | 4 - 5 | 590 | 72 | ||
| XM_007649292.1 | XP_007647482.1 | 4 - 5 | 590 | 72 | ||
| XM_007649293.1 | XP_007647483.1 | 5 - 6 | 459 | 72 | ||
| GenBank mRNA | JI888841.1 | - | 2 - 3 | 202 | 72 | |
| JP051559.1 | - | 2 - 3 | 202 | 72 | ||
More Information
Set Membership: |
|

Flt3lg
Genomic Map