Assay Details
| Gene Symbol: Hbegf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650976.1 | XP_007649166.1 | 3 - 4 | 648 | 69 | |
| GenBank mRNA | JI895907.1 | - | 2 - 3 | 252 | 69 | |
| JP060749.1 | - | 2 - 3 | 252 | 69 | ||
More Information
Set Membership: |
|

Hbegf
Genomic Map