Assay Details
| Gene Symbol: Bbs10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647105.1 | XP_007645295.1 | 1 - 2 | 226 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bbs10| Gene Symbol: Bbs10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647105.1 | XP_007645295.1 | 1 - 2 | 226 | 97 | |
Set Membership: |
|