Assay Details
Gene Symbol: Mif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508098.2 | XP_003508146.1 | 1 - 2 | 237 | 101 | |
GenBank mRNA | JI873677.1 | - | 1 - 2 | 218 | 101 | |
JP053046.1 | - | 1 - 2 | 218 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
