Assay Details
| Gene Symbol: Mif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508098.2 | XP_003508146.1 | 2 - 3 | 411 | 62 | |
| GenBank mRNA | JI873677.1 | - | 2 - 3 | 392 | 62 | |
| JP053046.1 | - | 2 - 3 | 392 | 62 | ||
More Information
Set Membership: |
|

Mif
Genomic Map