Assay Details
| Gene Symbol: Fam229b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503878.2 | XP_003503926.1 | 523 | 66 | ||
| GenBank mRNA | JI874308.1 | - | 499 | 66 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam229b| Gene Symbol: Fam229b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503878.2 | XP_003503926.1 | 523 | 66 | ||
| GenBank mRNA | JI874308.1 | - | 499 | 66 | ||
Set Membership: |
|