Assay Details
| Gene Symbol: Fam192a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503616.2 | XP_003503664.1 | 7 - 8 | 670 | 58 | |
| XM_007644209.1 | XP_007642399.1 | 7 - 8 | 853 | 58 | ||
| XM_007644210.1 | XP_007642400.1 | 7 - 8 | 954 | 58 | ||
| GenBank mRNA | JI886911.1 | - | 7 - 8 | 857 | 58 | |
| JP038642.1 | - | 7 - 8 | 857 | 58 | ||
More Information
Set Membership: |
|

Fam192a
Genomic Map