Assay Details
| Gene Symbol: Fam221a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507448.1 | XP_003507496.1 | 3 - 4 | 433 | 84 | |
| XM_007648039.1 | XP_007646229.1 | 3 - 4 | 433 | 84 | ||
| XM_007648040.1 | XP_007646230.1 | 2 - 3 | 259 | 84 | ||
More Information
Set Membership: |
|

Fam221a
Genomic Map