Assay Details
| Gene Symbol: Ahdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651966.1 | XP_007650156.1 | 1 - 2 | 4895 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ahdc1| Gene Symbol: Ahdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651966.1 | XP_007650156.1 | 1 - 2 | 4895 | 110 | |
Set Membership: |
|