Assay Details
| Gene Symbol: Vwa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509975.2 | XP_003510023.1 | 2 - 3 | 645 | 65 | |
| XM_007650640.1 | XP_007648830.1 | 2 - 3 | 645 | 65 | ||
More Information
Set Membership: |
|

Vwa1
Genomic Map