Assay Details
| Gene Symbol: Gid4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496350.2 | XP_003496398.1 | 5 - 6 | 416 | 84 | |
| GenBank mRNA | JI868112.1 | - | 4 - 5 | 361 | 84 | |
More Information
Set Membership: |
|

Gid4
Genomic Map