Assay Details
| Gene Symbol: Zcchc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510974.2 | XP_003511022.1 | 4 - 5 | 769 | 64 | |
| GenBank mRNA | JP042716.1 | - | 4 - 5 | 730 | 64 | |
More Information
Set Membership: |
|

Zcchc9
Genomic Map