Assay Details
Gene Symbol: Lamtor5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515839.2 | XP_003515887.2 | 1 - 2 | 158 | 68 | |
GenBank mRNA | JI881038.1 | - | 1 - 2 | 111 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
