Assay Details
| Gene Symbol: Tmem143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652678.1 | XP_007650868.1 | 5 - 6 | 740 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem143| Gene Symbol: Tmem143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652678.1 | XP_007650868.1 | 5 - 6 | 740 | 64 | |
Set Membership: |
|