Assay Details
| Gene Symbol: Limd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502976.2 | XP_003503024.2 | 2 - 3 | 180 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Limd2| Gene Symbol: Limd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502976.2 | XP_003503024.2 | 2 - 3 | 180 | 77 | |
Set Membership: |
|