Assay Details
| Gene Symbol: Dctn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645472.1 | XP_007643662.1 | 5 - 6 | 444 | 59 | |
| GenBank mRNA | JI882022.1 | - | 5 - 6 | 444 | 59 | |
More Information
Set Membership: |
|

Dctn3
Genomic Map