Assay Details
| Gene Symbol: Hsdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495122.2 | XP_003495170.1 | 4 - 5 | 840 | 59 | |
| XM_007608116.1 | XP_007606306.1 | 3 - 4 | 765 | 59 | ||
| XM_007608142.1 | XP_007606332.1 | 3 - 4 | 765 | 59 | ||
| GenBank mRNA | JI892737.1 | - | 1 - 2 | 190 | 59 | |
More Information
Set Membership: |
|

Hsdl1
Genomic Map